Learn More
Medchemexpress LLC 3-Aminophenylboronic acid | 30418-59-8 | MFCD00007755 | 99.9% | 136.94 g·mol⁻¹ | C6H8BNO2 | 1 G
SDP
Catalog No. 5000300001
Encompass_Preferred
3-Aminophenylboronic acid is an organoboron reagent supplied as a solid and used as a building block in organic synthesis and as a functional probe in biochemical assays. It is commonly used in bioconjugation workflows and can be attached to nanomaterials for selective glycoprotein sensing.
  • High purity (99.87%).
  • Solid, off-white to light brown physical form.
  • Molecular weight 136.94 g·mol⁻¹.
  • Useful as a synthetic building block and reagent for bioconjugation.
  • Suits functionalization of nanomaterials for selective glycoprotein detection.
  • Available in multiple package sizes, including 1 g.
Catalog No. 50-003-00001 Supplier Medchemexpress LLC Supplier No. HYW0176041G
3-Aminophenylboronic acid is an organoboron reagent supplied as a solid and used as a building block in organic synthesis and as a functional probe in biochemical assays. It is commonly used in bioconjugation workflows and can be attached to nanomaterials for selective glycoprotein sensing.
  • High purity (99.87%).
  • Solid, off-white to light brown physical form.
  • Molecular weight 136.94 g·mol⁻¹.
  • Useful as a synthetic building block and reagent for bioconjugation.
  • Suits functionalization of nanomaterials for selective glycoprotein detection.
  • Available in multiple package sizes, including 1 g.