Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-aminophenylboronic acid | 30418-59-8 | MFCD00007755 | 99.9% | 136.94 g/mol | C6H8BNO2 | 1 ML
SDP

Catalog No. 5000301449
Encompass_Preferred

3-Aminophenylboronic acid is an organoboron research reagent used in biochemical assays and organic synthesis. It is employed to functionalize nanomaterials for selective glycoprotein detection and serves as a versatile building block in cross-coupling and labeling reactions. The product is available in both ready-to-use solution and solid formats to support varied laboratory workflows.

  • High purity (99.87%).
  • Molecular weight 136.94 g/mol.
  • Chemical formula C6H8BNO2.
  • Available as a 10 mM solution in DMSO (1 mL) and multiple solid pack sizes.
  • Suitable for glycoprotein detection, biochemical assays, and synthetic applications.

Catalog No. 50-003-01449 Supplier Medchemexpress LLC Supplier No. HYW01760410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Aminophenylboronic acid is an organoboron research reagent used in biochemical assays and organic synthesis. It is employed to functionalize nanomaterials for selective glycoprotein detection and serves as a versatile building block in cross-coupling and labeling reactions. The product is available in both ready-to-use solution and solid formats to support varied laboratory workflows.

  • High purity (99.87%).
  • Molecular weight 136.94 g/mol.
  • Chemical formula C6H8BNO2.
  • Available as a 10 mM solution in DMSO (1 mL) and multiple solid pack sizes.
  • Suitable for glycoprotein detection, biochemical assays, and synthetic applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.