Learn More
Medchemexpress LLC 3-aminophenylboronic acid | 30418-59-8 | MFCD00007755 | 99.9% | 136.94 g/mol | C6H8BNO2 | 25 G
SDP
Catalog No. 5000302709
Encompass_Preferred
3-Aminophenylboronic acid is an organoboron reagent used as a building block in organic synthesis and biochemical research. It participates in cross-coupling reactions and enables boronic-acid-based functionalization of materials, for example in nanoparticle conjugation and glycoprotein sensing. Supplied as a high-purity solid, it is suitable for analytical and preparative laboratory work.
  • High purity suitable for research.
  • Useful in Suzuki-Miyaura and other cross-coupling reactions.
  • Reacts with diols for reversible covalent binding to saccharides.
  • Enables functionalization of nanomaterials and sensors.
  • Available in multiple pack sizes for lab-scale and bulk use.
Catalog No. 50-003-02709 Supplier Medchemexpress LLC Supplier No. 25GHYW017604
3-Aminophenylboronic acid is an organoboron reagent used as a building block in organic synthesis and biochemical research. It participates in cross-coupling reactions and enables boronic-acid-based functionalization of materials, for example in nanoparticle conjugation and glycoprotein sensing. Supplied as a high-purity solid, it is suitable for analytical and preparative laboratory work.
  • High purity suitable for research.
  • Useful in Suzuki-Miyaura and other cross-coupling reactions.
  • Reacts with diols for reversible covalent binding to saccharides.
  • Enables functionalization of nanomaterials and sensors.
  • Available in multiple pack sizes for lab-scale and bulk use.