Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-aminophenylboronic acid | 30418-59-8 | MFCD00007755 | 99.9% | 136.94 g·mol⁻¹ | C6H8BNO2 | 5 G
SDP

Catalog No. 5000300499
Encompass_Preferred

3-Aminophenylboronic acid is an organoboron reagent used in biochemical assays and organic synthesis. It engages in boronate-diol chemistry for selective binding of glycoproteins and can be functionalized on nanomaterials for sensing applications. The compound is supplied as a high-purity solid suitable for laboratory research and conjugation workflows.

  • High purity suitable for analytical and research use.
  • Binds selectively to cis-diols for glycoprotein recognition.
  • Soluble in DMSO for convenient sample preparation.
  • Supplied as a solid powder in lab-scale packaging.
  • Molecular formula C6H8BNO2; molecular weight 136.94 g·mol⁻¹.

Catalog No. 50-003-00499 Supplier Medchemexpress LLC Supplier No. HYW0176045G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Aminophenylboronic acid is an organoboron reagent used in biochemical assays and organic synthesis. It engages in boronate-diol chemistry for selective binding of glycoproteins and can be functionalized on nanomaterials for sensing applications. The compound is supplied as a high-purity solid suitable for laboratory research and conjugation workflows.

  • High purity suitable for analytical and research use.
  • Binds selectively to cis-diols for glycoprotein recognition.
  • Soluble in DMSO for convenient sample preparation.
  • Supplied as a solid powder in lab-scale packaging.
  • Molecular formula C6H8BNO2; molecular weight 136.94 g·mol⁻¹.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.