Learn More
Medchemexpress LLC 3-aminophenylboronic acid | 30418-59-8 | MFCD00007755 | 99.9% | 136.94 g/mol | C6H8BNO2 | 500 G
SDP
Catalog No. 5000300207
Encompass_Preferred
3-Aminophenylboronic acid is an organoboron reagent used in organic synthesis and biochemical research. It serves as a building block for cross-coupling chemistry and can be functionalized for biosensing applications, such as glycoprotein recognition when attached to nanomaterials. The compound is supplied at high purity and is suitable for analytical, preparative, and material science workflows.
  • High purity suitable for analytical and synthetic applications.
  • Serves as a boronic acid building block for cross-coupling reactions.
  • Contains a reactive amino group for derivatization and bioconjugation.
  • Applicable to sensor development for selective glycoprotein detection.
  • Available in packaging sizes that support scale-up and bulk use.
Catalog No. 50-003-00207 Supplier Medchemexpress LLC Supplier No. HYW017604500G
3-Aminophenylboronic acid is an organoboron reagent used in organic synthesis and biochemical research. It serves as a building block for cross-coupling chemistry and can be functionalized for biosensing applications, such as glycoprotein recognition when attached to nanomaterials. The compound is supplied at high purity and is suitable for analytical, preparative, and material science workflows.
  • High purity suitable for analytical and synthetic applications.
  • Serves as a boronic acid building block for cross-coupling reactions.
  • Contains a reactive amino group for derivatization and bioconjugation.
  • Applicable to sensor development for selective glycoprotein detection.
  • Available in packaging sizes that support scale-up and bulk use.