Learn More
Medchemexpress LLC 3-Aminophenylboronic acid monohydrate | 206658-89-1 | 154.96 | 10 G
SDP
Catalog No. 5000449721
Encompass_Preferred
3-Aminophenylboronic acid monohydrate is a biochemical reagent useful as a biological material or organic compound for life science related research. It is intended for research use only and not for sale to patients.
  • Reagent in the preparation of Suzuki-Miyaura cross-coupling
  • Used for Gram-positive anti-virulence drugs and inhibitors of Streptococcus agalactiae Stk1
  • Regioisomer of Zaleplon, a sedative
  • Amphiphilic random glycopolymer that forms nanoparticles
  • Chemomechanical polymer that expands and contracts in blood plasma with high glucose selectivity
  • Reagent in the preparation of nonbenzodiazepine hypnotic agents
Catalog No. 50-004-49721 Supplier Medchemexpress LLC Supplier No. HYW00089710G
3-Aminophenylboronic acid monohydrate is a biochemical reagent useful as a biological material or organic compound for life science related research. It is intended for research use only and not for sale to patients.
  • Reagent in the preparation of Suzuki-Miyaura cross-coupling
  • Used for Gram-positive anti-virulence drugs and inhibitors of Streptococcus agalactiae Stk1
  • Regioisomer of Zaleplon, a sedative
  • Amphiphilic random glycopolymer that forms nanoparticles
  • Chemomechanical polymer that expands and contracts in blood plasma with high glucose selectivity
  • Reagent in the preparation of nonbenzodiazepine hypnotic agents