Learn More
Medchemexpress LLC HY-107829 100mg Medchemexpress, 3-Aminopropionitrile fumarate (2:1) CAS:2079-89-2 Purity:>98%
SDP
Catalog No. 502080723
Encompass_Preferred
Medchemexpress, HY-107829 100mg 3-Aminopropionitrile fumarate (2:1) CAS:2079-89-2 3-Aminopropionitrile fumarate (2:1) is a lathyrogen which inhibits crosslinking of collagen. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0723 Supplier Medchemexpress LLC Supplier No. HY107829100MG
Medchemexpress, HY-107829 100mg 3-Aminopropionitrile fumarate (2:1) CAS:2079-89-2 3-Aminopropionitrile fumarate (2:1) is a lathyrogen which inhibits crosslinking of collagen. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.