Learn More
Apexbio Technology LLC 3-Aminopropionitrile fumarate (2:1) 2079-89-2 500mg
SDP
Catalog No. 5000094915
Encompass_Preferred
3-Aminopropionitrile fumarate (2 1) (CAS 2079-89-2) is a small molecule derivative originally identified from Apocynaceae species It acts by inhibiting collagen cross-linking through interference with enzymatic pathways responsible for covalent bonding among collagen molecules Modulation of enzyme activity and signaling cascades by this compound leads to altered collagen stability within biological tissues Owing to its capacity to disrupt collagen maturation 3-Aminopropionitrile fumarate (2 1) serves as a valuable tool in studies of fibrotic disorders and related metabolic processes where aberrant extracellular matrix remodeling is implicated
Catalog No. 50-000-94915 Supplier Apexbio Technology LLC Supplier No. A4634500
3-Aminopropionitrile fumarate (2 1) (CAS 2079-89-2) is a small molecule derivative originally identified from Apocynaceae species It acts by inhibiting collagen cross-linking through interference with enzymatic pathways responsible for covalent bonding among collagen molecules Modulation of enzyme activity and signaling cascades by this compound leads to altered collagen stability within biological tissues Owing to its capacity to disrupt collagen maturation 3-Aminopropionitrile fumarate (2 1) serves as a valuable tool in studies of fibrotic disorders and related metabolic processes where aberrant extracellular matrix remodeling is implicated