Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-aminopropionitrile fumarate (2:1) | 2079-89-2 | MFCD00013142 | 99.9% | 256.26 g/mol | C10H16N4O4 | 500 MG
SDP

Catalog No. 5000242354
Encompass_Preferred

3-aminopropionitrile fumarate (2:1) is a research-grade lathyrogen that inhibits collagen crosslinking. Supplied as a white to off-white solid, it is used in studies of extracellular matrix remodeling, fibrosis, and collagen-related biochemical assays. Available in multiple sizes for laboratory use.

  • Inhibits enzymatic crosslinking of collagen.
  • High purity, 99.9%.
  • Molecular weight 256.26 g/mol; formula C10H16N4O4.
  • White to off-white solid.
  • Store at 4°C in a sealed container away from moisture.
  • Supplied in multiple sizes including 500 mg for bench-scale experiments.

Catalog No. 50-002-42354 Supplier Medchemexpress LLC Supplier No. HY107829500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-aminopropionitrile fumarate (2:1) is a research-grade lathyrogen that inhibits collagen crosslinking. Supplied as a white to off-white solid, it is used in studies of extracellular matrix remodeling, fibrosis, and collagen-related biochemical assays. Available in multiple sizes for laboratory use.

  • Inhibits enzymatic crosslinking of collagen.
  • High purity, 99.9%.
  • Molecular weight 256.26 g/mol; formula C10H16N4O4.
  • White to off-white solid.
  • Store at 4°C in a sealed container away from moisture.
  • Supplied in multiple sizes including 500 mg for bench-scale experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.