Learn More
Medchemexpress LLC 3-aminopropionitrile fumarate (2:1) | 2079-89-2 | MFCD00078321 | 99.9% | 256.26 | C10H16N4O4 | 5 G
SDP
Catalog No. 5000242266
Encompass_Preferred
3-Aminopropionitrile fumarate (2:1) is the fumarate salt of 3-aminopropionitrile, a lathyrogen that inhibits collagen crosslinking. This high-purity research reagent is used to study extracellular matrix remodeling, lysyl oxidase-related pathways, and mechanisms of collagen maturation in biochemical and cell biology experiments.
  • High purity (99.9%) suitable for biochemical and cellular assays.
  • Fumarate salt form with defined 2:1 stoichiometry for reproducible results.
  • Acts as a lysyl oxidase inhibitor to study collagen crosslinking processes.
  • Available in laboratory-friendly pack sizes, including 5 g.
Catalog No. 50-002-42266 Supplier Medchemexpress LLC Supplier No. HY1078295G
3-Aminopropionitrile fumarate (2:1) is the fumarate salt of 3-aminopropionitrile, a lathyrogen that inhibits collagen crosslinking. This high-purity research reagent is used to study extracellular matrix remodeling, lysyl oxidase-related pathways, and mechanisms of collagen maturation in biochemical and cell biology experiments.
  • High purity (99.9%) suitable for biochemical and cellular assays.
  • Fumarate salt form with defined 2:1 stoichiometry for reproducible results.
  • Acts as a lysyl oxidase inhibitor to study collagen crosslinking processes.
  • Available in laboratory-friendly pack sizes, including 5 g.