Learn More
Medchemexpress LLC 3-aminopropionitrile fumarate (2:1) | 2079-89-2 | MFCD00078321 | 99.9% | 256.26 | C4H4O4·2C3H6N2 | 1 G
SDP
Catalog No. 5000240269
Encompass_Preferred
3-Aminopropionitrile fumarate (2:1) is a lathyrogen used in research to inhibit crosslinking of collagen. Supplied as a solid salt and as an aqueous solution, it is intended for research use only and should be stored sealed at 4°C; solutions require low-temperature storage for extended stability.
  • Inhibits crosslinking of collagen.
  • Available as solid (100 mg-50 g) and as 10 mM aqueous solution.
  • High purity (≈99.9%).
  • Molecular weight 256.26 g/mol.
  • Store sealed at 4°C; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • For research use only; not for human or clinical applications.
Catalog No. 50-002-40269 Supplier Medchemexpress LLC Supplier No. HY1078291G
3-Aminopropionitrile fumarate (2:1) is a lathyrogen used in research to inhibit crosslinking of collagen. Supplied as a solid salt and as an aqueous solution, it is intended for research use only and should be stored sealed at 4°C; solutions require low-temperature storage for extended stability.
  • Inhibits crosslinking of collagen.
  • Available as solid (100 mg-50 g) and as 10 mM aqueous solution.
  • High purity (≈99.9%).
  • Molecular weight 256.26 g/mol.
  • Store sealed at 4°C; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • For research use only; not for human or clinical applications.