Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-aminopropionitrile fumarate (2:1) | 2079-89-2 | MFCD00078321 | 99.9% | 256.26 g·mol⁻¹ | C4H4O4·2C3H6N2 | 1 ML
SDP

Catalog No. 5000242795
Encompass_Preferred

3-Aminopropionitrile fumarate (2:1) is the fumarate salt of 3-aminopropionitrile and acts as a lathyrogen by inhibiting lysyl oxidase (LOX), preventing collagen crosslinking in research applications. It is supplied as a white to off-white solid and is commonly available pre-formulated as a 10 mM solution (1 mL) for in vitro use. Reported purity is 99.94%.

  • Inhibits collagen crosslinking via lysyl oxidase inhibition.
  • Available as solid or ready-to-use 10 mM solution, 1 mL.
  • High reported purity (99.94%).
  • Molecular weight 256.26 g·mol⁻¹ and solid appearance.
  • Recommended storage: solid at 4°C; in solvent, -80°C (up to 6 months) or -20°C (short term).

Catalog No. 50-002-42795 Supplier Medchemexpress LLC Supplier No. HY10782910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Aminopropionitrile fumarate (2:1) is the fumarate salt of 3-aminopropionitrile and acts as a lathyrogen by inhibiting lysyl oxidase (LOX), preventing collagen crosslinking in research applications. It is supplied as a white to off-white solid and is commonly available pre-formulated as a 10 mM solution (1 mL) for in vitro use. Reported purity is 99.94%.

  • Inhibits collagen crosslinking via lysyl oxidase inhibition.
  • Available as solid or ready-to-use 10 mM solution, 1 mL.
  • High reported purity (99.94%).
  • Molecular weight 256.26 g·mol⁻¹ and solid appearance.
  • Recommended storage: solid at 4°C; in solvent, -80°C (up to 6 months) or -20°C (short term).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.