Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminopropionitrile Fumarate 98.0+%, TCI America™
SDP

Catalog No. A079625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A079625G 25 g
A0796500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A079625G Supplier TCI America Supplier No. A079625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2079-89-2
Molecular Formula C3H6N2
MDL Number MFCD00078321

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 256.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminopropionitrile Fumarate
RTECSNumber UG0700000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.