Learn More
Sigma Aldrich 3-Aminopropyl-Functionalized Silica Gel
Catalog No. 36425850G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-425-850G 50 g
36-425-8250G 250 g
36-425-810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 36-425-850G Supplier Sigma Aldrich Supplier No. 36425850G
Only null left
Add to Cart
Add to Cart
Warning
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
The product is silica gel functionalized with amino groups. The product (3-Aminopropyl-functionalized silica gel) is 9% functionalized and has 1mmol/g of NH2 loading (extent of labelling). It participates as catalyst in the Knoevenagel condensation reactions.3-Aminopropyl-functionalized silica gel may be used in the following studies:For the separation of polychlorinated biphenyls (PCBs) fro mLipids by combined supercritical fluid extraction and separation.Preparation of the tin complex immobilized on silica gel, a novel, green, highly efficient and heterogeneous reusable catalyst.Preparation of silica gel bearing isonicotinamide groups.Synthesis of a novel catalytic system including Pd(OAc)2 attached to a bis(oxazoline) ligand that is covalently bonded to 3-mercaptopropyl-functionalized silica gel.Useful as a heterogeneous, reusable catalyst for the Knoevenagel condensation.For low- and medium-pressure reversed-phase liquid chromatography.
Specifications
MDL Number MFCD00132846
Quantity 50 g
Biological Activity Respiratory System