Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (3-aminopropyl)(n-butyl)phosphinic acid | 123690-78-8 | 98.0% | 10 MG
SDP

Catalog No. 5000230559
Encompass_Preferred

CGP 36742 (SGS-742) is an orally active, selective GABAB receptor antagonist supplied as a solid research reagent and analytical standard for preclinical neuroscience and pharmacology studies.

  • Purity 98.0%.
  • White to light yellow solid.
  • Molecular formula C7H18NO2P and molecular weight 179.20 g·mol⁻¹.
  • Supplied in 10 MG solid format; other sizes and solution formats are available.
  • Used in GABAB receptor pharmacology and cognitive performance research.

Catalog No. 50-002-30559 Supplier Medchemexpress LLC Supplier No. HY12159910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CGP 36742 (SGS-742) is an orally active, selective GABAB receptor antagonist supplied as a solid research reagent and analytical standard for preclinical neuroscience and pharmacology studies.

  • Purity 98.0%.
  • White to light yellow solid.
  • Molecular formula C7H18NO2P and molecular weight 179.20 g·mol⁻¹.
  • Supplied in 10 MG solid format; other sizes and solution formats are available.
  • Used in GABAB receptor pharmacology and cognitive performance research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.