Learn More
Medchemexpress LLC (3-aminopropyl)(n-butyl)phosphinic acid | 123690-78-8 | 98.0% | 50 MG
SDP
Catalog No. 5000232052
Encompass_Preferred
CGP 36742 (SGS-742) is an orally active, blood-brain barrier-penetrant, selective GABAB receptor antagonist (reported IC50 ≈ 36 μM) supplied as a research-grade compound for in vitro and in vivo neuroscience studies, including depression and cognition research.
  • Selective GABAB receptor antagonist with reported IC50 ≈ 36 μM.
  • Penetrates the blood-brain barrier, suitable for CNS pharmacology studies.
  • Provided as a solid research reagent in small milligram pack sizes.
  • Commonly used in behavioral pharmacology and receptor binding studies related to depression and cognition.
  • Reported purity typically around 98.0%, varying by supplier and lot.
Catalog No. 50-002-32052 Supplier Medchemexpress LLC Supplier No. HY12159950MG
CGP 36742 (SGS-742) is an orally active, blood-brain barrier-penetrant, selective GABAB receptor antagonist (reported IC50 ≈ 36 μM) supplied as a research-grade compound for in vitro and in vivo neuroscience studies, including depression and cognition research.
  • Selective GABAB receptor antagonist with reported IC50 ≈ 36 μM.
  • Penetrates the blood-brain barrier, suitable for CNS pharmacology studies.
  • Provided as a solid research reagent in small milligram pack sizes.
  • Commonly used in behavioral pharmacology and receptor binding studies related to depression and cognition.
  • Reported purity typically around 98.0%, varying by supplier and lot.