Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (3-Aminopropyl)triethoxysilane

Catalog No. 70649320ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-649-320ML 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-649-320ML Supplier Sigma Aldrich Supplier No. 70649320ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Atomic number of base material: 14 SiliconPrecursors Packaged for Depositions Systems

Specifications

CAS 919-30-2
Density 0.946 g/mL (at 25°C)
Boiling Point 217°C (760 mmHg)
Molecular Formula C9H23NO3Si
Linear Formula H2N(CH2)3 Si(OC2H5)3
Quantity 20 mL
MDL Number MFCD00008207
Synonym 3-Triethoxysilylpropylamine; APTES; APTS
Molecular Weight (g/mol) 221.37
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.