Learn More
Medchemexpress LLC (3-aminopropyl)trimethoxysilane | 13822-56-5 | MFCD00008206 | >96.0% | 179.29 g·mol⁻¹ | C6H17NO3Si | 100 G
SDP
Catalog No. 5000301876
Encompass_Preferred
3-Aminopropyltrimethoxysilane is an organosilane coupling agent that introduces amino-functional silane groups onto inorganic surfaces. It is used for silanization of glass and silica, surface modification, adhesion promotion, and as a precursor in materials science, coatings, and biosensor surface chemistry.
  • Provides amino functional groups for covalent coupling to surfaces.
  • Enhances adhesion between organic polymers and inorganic substrates.
  • Commonly used for silanization of glass, silica, and oxide surfaces.
  • Colorless to almost colorless clear liquid with typical purity >96%.
  • Suitable for coatings, composite coupling, and surface functionalization applications.
Catalog No. 50-003-01876 Supplier Medchemexpress LLC Supplier No. HYW036410100G
3-Aminopropyltrimethoxysilane is an organosilane coupling agent that introduces amino-functional silane groups onto inorganic surfaces. It is used for silanization of glass and silica, surface modification, adhesion promotion, and as a precursor in materials science, coatings, and biosensor surface chemistry.
  • Provides amino functional groups for covalent coupling to surfaces.
  • Enhances adhesion between organic polymers and inorganic substrates.
  • Commonly used for silanization of glass, silica, and oxide surfaces.
  • Colorless to almost colorless clear liquid with typical purity >96%.
  • Suitable for coatings, composite coupling, and surface functionalization applications.