Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (3-Aminopropyl)trimethoxysilane

Catalog No. 281778500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-177-8500ML 500 mL
28-177-85ML 5 mL
28-177-8100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 28-177-8500ML Supplier Sigma Aldrich Supplier No. 281778500ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(3-Aminopropyl)trimethoxysilane(APTMS) is an amino-silane that is mainly used as a silane coupling agent for the surface modification of a variety of nanomaterials. APTMS based films form a thermally stable layer on different substrates.APTMS is used for the synthesis of gold nano-bipyramids which can be used as substrates for C-reactive protein (CRP) antibodies that were detected by localized surface plasmon resonance(LSPR). Mesoporous silica matrix can be incorporated with APTMS for the removal of chromium (Cr) from waste water. It can also be used to silanize magnetic iron oxide nanoparticles(MIONPs) for the separation of cross-linked enzyme aggregations (CLEAs) from the reaction medium. It may be coated on TiO2 to improve the power conversion efficiency (PCE) as a critical parameter to assess the overall performance of heterojunction perovskite solar cells (PSCs).

Specifications

CAS 13822-56-5
Density 1.027 g/mL (at 25°C)
Boiling Point 91°C to 92°C (15 mmHg)
Molecular Formula C6H17NO3Si
Refractive Index n20/D 1.424 (literature)
Linear Formula H2N(CH2)3 Si(OCH3)3
Quantity 500 mL
MDL Number MFCD00008206
Synonym 3-(Trimethoxysilyl)propylamine
Molecular Weight (g/mol) 179.29
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.