Learn More
Gelest Inc 3-Aminopropyldimethylethoxysilane, 18306-79-1, MFCD00053947, 5g
Supplier:  Gelest Inc SIA0603.05G
Encompass
This item has a minimum qty of 7 per supplier requirements.
Molecular Formual: C7H19NOSI, Molecular Weight: 161.32, Boiling Point: 78-9 / 24
Catalog No. NC9531099