Learn More
Medchemexpress LLC 3-Aminopropylphospho 250mg
SDP
Catalog No. 5000205308
Encompass_Preferred
3-Aminopropylphosphonic acid ((3-Aminopropyl)phosphonic acid) is a phosphonic acid analogue of GABA and a GABAB receptor agonist[1][2]
Catalog No. 50-002-05308 Supplier Medchemexpress LLC Supplier No. 250MGHYW105694
3-Aminopropylphosphonic acid ((3-Aminopropyl)phosphonic acid) is a phosphonic acid analogue of GABA and a GABAB receptor agonist[1][2]