Learn More
Medchemexpress LLC 3-Aminopropyltriethoxysilane | 919-30-2 | 99.8% | 100 G
SDP
Catalog No. 5000410827
Encompass_Preferred
3-Aminopropyltriethoxysilane (APTES) functions as a strong adhesive for immobilizing biomolecules such as antibodies and enzymes onto silicon and its derivatives like silicon nitride (Si3N4). It also serves as a spacer, enhancing the spatial freedom of biomolecules during immobilization, which leads to higher specific activity. This compound can establish a more stable, sensitive, and highly biocompatible bioanalytical platform by immobilizing biomolecules onto various solid materials, electrode materials, nanomaterials, and nanocomposites.
  • Acts as a strong glue to immobilize biomolecules (e.g., antibodies and enzymes) to silicon and silicon derivatives.
  • Functions as a spacer, providing biomolecules with more spatial freedom for higher specific activity.
  • Can form a more stable, sensitive, and highly biocompatible bioanalytical platform.
Catalog No. 50-004-10827 Supplier Medchemexpress LLC Supplier No. HYD0175100G
3-Aminopropyltriethoxysilane (APTES) functions as a strong adhesive for immobilizing biomolecules such as antibodies and enzymes onto silicon and its derivatives like silicon nitride (Si3N4). It also serves as a spacer, enhancing the spatial freedom of biomolecules during immobilization, which leads to higher specific activity. This compound can establish a more stable, sensitive, and highly biocompatible bioanalytical platform by immobilizing biomolecules onto various solid materials, electrode materials, nanomaterials, and nanocomposites.
  • Acts as a strong glue to immobilize biomolecules (e.g., antibodies and enzymes) to silicon and silicon derivatives.
  • Functions as a spacer, providing biomolecules with more spatial freedom for higher specific activity.
  • Can form a more stable, sensitive, and highly biocompatible bioanalytical platform.