Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminopyridazine, 95%, Thermo Scientific™

Catalog No. AC454172500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454172500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC454172500 Supplier Thermo Scientific Chemicals Supplier No. 454172500

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5469-70-5
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.1
InChI Key LETVJWLLIMJADE-UHFFFAOYSA-N
Synonym 3-aminopyridazine, 3-pyridazinamine, aminopyridazine, pyridazin-3-ylamine, 3-amino-1,2-diazine, pyridazine-3-ylamine, pyridazineamine, pyridazyl amine, imino-pyridazine, 3-pyridazinamin
PubChem CID 230373
IUPAC Name pyridazin-3-amine
SMILES C1=CC(=NN=C1)N

Specifications

Melting Point 168.0°C to 172.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 95.1
Percent Purity 95%
Chemical Name or Material 3-Aminopyridazine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.