Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminopyrrolidine 99.0+%, TCI America™
SDP

Catalog No. A11045G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11045G 5 g
A110425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11045G Supplier TCI America Supplier No. A11045G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 79286-79-6
Molecular Formula C4H10N2
Molecular Weight (g/mol) 86.138
MDL Number MFCD00059018
InChI Key NGXSWUFDCSEIOO-UHFFFAOYSA-N
PubChem CID 164401
IUPAC Name pyrrolidin-3-amine
SMILES C1CNCC1N

Specifications

Color Yellow
Quantity 5 g
UN Number 2735
Formula Weight 86.14
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Aminopyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.