Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Aminosalicylic acid | 570-23-0 | MFCD00010299 | 99.1% | 153.14 | C7H7NO3 | 1 G
SDP

Catalog No. 5000283197
Encompass_Preferred

3-Aminosalicylic acid is a small-molecule impurity formed during the synthesis of 5-aminosalicylic acid and provided for research and analytical applications. The compound has formula C7H7NO3 and a molecular weight of 153.14 g/mol. It is supplied at high purity and requires storage protected from light under inert conditions.

  • High purity (99.06%) suitable for analytical reference.
  • Chemical formula C7H7NO3 and molecular weight 153.14 g/mol.
  • Stored at 4°C and protected from light; in solvent, store at -80°C (long term) or -20°C (short term).
  • Intended for research and analytical use; not for human therapeutic use.
  • Supplied as a 1 g solid.

Catalog No. 50-002-83197 Supplier Medchemexpress LLC Supplier No. HYW2060161G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

3-Aminosalicylic acid is a small-molecule impurity formed during the synthesis of 5-aminosalicylic acid and provided for research and analytical applications. The compound has formula C7H7NO3 and a molecular weight of 153.14 g/mol. It is supplied at high purity and requires storage protected from light under inert conditions.

  • High purity (99.06%) suitable for analytical reference.
  • Chemical formula C7H7NO3 and molecular weight 153.14 g/mol.
  • Stored at 4°C and protected from light; in solvent, store at -80°C (long term) or -20°C (short term).
  • Intended for research and analytical use; not for human therapeutic use.
  • Supplied as a 1 g solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.