Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminosalicylic acid, 97%

Catalog No. p-58213
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC225260500 50 mg Glass bottle
AC225262500 250 mg Glass bottle
AC225260010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC225260500 Supplier Thermo Scientific Chemicals Supplier No. 225260500
Only null left
Add to Cart

CAS: 570-23-0 | C7H7NO3 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 570-23-0
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00010299
InChI Key IQGMRVWUTCYCST-UHFFFAOYSA-N
Synonym 3-aminosalicylic acid, 3-amino salicylic acid, benzoic acid, 3-amino-2-hydroxy, 3-amino-2-hydroxy-benzoic acid, 2-amino-6-carboxyphenol, salicylic acid, 3-amino, 3-carboxy-2-hydroxyaniline, 3 or 5-aminosalicylic acid, benzoic acid, amino-2-hydroxy
PubChem CID 68443
IUPAC Name 3-amino-2-hydroxybenzoic acid
SMILES C1=CC(=C(C(=C1)N)O)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 240.0°C
Color Beige to Gray-Green
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 50 mg
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetone, slightly soluble in ethanol
Formula Weight 153.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Aminosalicylic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-328-0
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.