Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminotetrahydro-1H-1lambda∼6∼-thiophene-1,1-dione hydrochloride , Tech., Thermo Scientific™

Catalog No. RJC02399CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RJC02399CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RJC02399CB Supplier Thermo Scientific Chemicals Supplier No. RJC02399CB
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 51642-03-6
Molecular Formula C4H10ClNO2S
Molecular Weight (g/mol) 171.64
MDL Number MFCD00456584
InChI Key MGZQMSFXPSKBDY-UHFFFAOYNA-N
Synonym 3-aminotetrahydrothiophene 1,1-dioxide hydrochloride, 1,1-dioxidotetrahydrothien-3-ylamine hydrochloride, tetrahydro-3-thiophenamine 1,1-dioxide hydrochloride, 1,1-dioxo-tetrahydrothiophen-3-ylamine, hcl, 3-amino-1??-thiolane-1,1-dione hydrochloride, 1,1-dioxidotetrahydro-3-thienyl amine hydrochloride, 3-aminotetrahydrothiopene 1,1-dioxide hydrochloride, tetrahydrothiophen-3-amine 1,1-dioxide hydrochloride, 3-thiophenamine, tetrahydro-, 1,1-dioxide, hydrochloride, 1,1-dioxothiolan-3-amine hydrochloride
PubChem CID 2795201
SMILES [H+].[Cl-].NC1CCS(=O)(=O)C1

Specifications

Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name hydrogen 3-amino-1λ⁶-thiolane-1,1-dione chloride
Formula Weight 171.65
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-Aminotetrahydrothiopene 1, 1-dioxide hydrochloride
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.