Learn More
Medchemexpress LLC 3α-Angeloyloxypterokaurene L3 | 79406-11-4 | 416.55 | 5 MG
SDP
Catalog No. 5000415055
Encompass_Preferred
3α-Angeloyloxypterokaurene L3 is a Diterpenoids product isolated from the herbs of *Wedelia trilobata*. It is a white to off-white solid, primarily classified under Terpenoids and Diterpenoids, with initial sources from plants such as Leguminosae (*Senna siamea*) and Compositae. This compound is intended for research purposes only.
  • Isolated from *Wedelia trilobata* and other plant sources.
  • Diterpenoid product.
  • Appears as a white to off-white solid.
  • High purity (98.0%).
  • Specific storage conditions (-20°C, sealed, away from moisture and light) ensure product stability.
Catalog No. 50-004-15055 Supplier Medchemexpress LLC Supplier No. HYN18115MG
3α-Angeloyloxypterokaurene L3 is a Diterpenoids product isolated from the herbs of *Wedelia trilobata*. It is a white to off-white solid, primarily classified under Terpenoids and Diterpenoids, with initial sources from plants such as Leguminosae (*Senna siamea*) and Compositae. This compound is intended for research purposes only.
  • Isolated from *Wedelia trilobata* and other plant sources.
  • Diterpenoid product.
  • Appears as a white to off-white solid.
  • High purity (98.0%).
  • Specific storage conditions (-20°C, sealed, away from moisture and light) ensure product stability.