Learn More
Enzo Life Sciences 3-ATA (5mg). CAS: 129821-08-5
SDP
Catalog No. 502008819
Encompass_Preferred
Specific CDK4 inhibitor (IC50=3.1µM) inducing growth inhibition of p16-altered tumors. Alternative name: 3-Amino-9-thio(10H)-acridone. Purity: ≥95%. Appearance: White to reddish brown powder. Solubility: Soluble in DMSO or dimethyl formamide. Long Term Storage: Ambient.
Catalog No. 50-200-8819 Supplier Enzo Life Sciences Supplier No. ALX350273M005
Specific CDK4 inhibitor (IC50=3.1µM) inducing growth inhibition of p16-altered tumors. Alternative name: 3-Amino-9-thio(10H)-acridone. Purity: ≥95%. Appearance: White to reddish brown powder. Solubility: Soluble in DMSO or dimethyl formamide. Long Term Storage: Ambient.