Learn More
World Precision Instrument Micrometer Slide Micromanipulator
SDP
Catalog No. NC0725957
Encompass
Micrometer Slide Micromanipulator
The M325 three-axis man­u­al mi­cro­ma­n­ip­u­la­tor is built of preci­sion mi­crome­ter-ac­tu­at­ed lin­ear slides. Each slide is com­prised of a large microme­ter head and a spring-return linear slide. The mi­cro­ma­n­ip­u­la­tor was designed to min­i­mize wear in the moving components to achieve a long op­er­a­tion­al life without the necessity for fre­quent main­te­nance. The mi­crome­ter head is grad­u­at­ed in 10 µm steps which enable re­peat­able po­si­tion­ing to an ac­cu­ra­cy of ± 2 µm.
A unique spring return mechanism is used to trans­mit move­ment of the microme­ter spin­dle to the slide car­riage, eliminating back­lash, lost motion and re­duc­ing thread wear. Each lin­ear slide utilizes ball bear­ings which en­able the M325 to carry loads of up to 1 kg.
The tool holder can clamp onto tools with shaft di­am­e­ters of 3.0 mm to 12.7mm and allows rotation around two axes.
Catalog No. NC0725957 Supplier World Precision Instrument Supplier No. M325
Micrometer Slide Micromanipulator
The M325 three-axis man­u­al mi­cro­ma­n­ip­u­la­tor is built of preci­sion mi­crome­ter-ac­tu­at­ed lin­ear slides. Each slide is com­prised of a large microme­ter head and a spring-return linear slide. The mi­cro­ma­n­ip­u­la­tor was designed to min­i­mize wear in the moving components to achieve a long op­er­a­tion­al life without the necessity for fre­quent main­te­nance. The mi­crome­ter head is grad­u­at­ed in 10 µm steps which enable re­peat­able po­si­tion­ing to an ac­cu­ra­cy of ± 2 µm.
A unique spring return mechanism is used to trans­mit move­ment of the microme­ter spin­dle to the slide car­riage, eliminating back­lash, lost motion and re­duc­ing thread wear. Each lin­ear slide utilizes ball bear­ings which en­able the M325 to carry loads of up to 1 kg.
The tool holder can clamp onto tools with shaft di­am­e­ters of 3.0 mm to 12.7mm and allows rotation around two axes.