Learn More
Medchemexpress LLC 3-azaspiro[5.5]undecane hydrochloride | 1125-01-5 | MFCD02093801 | 97.0% | 189.73 g·mol⁻¹ | C10H20ClN | 250 MG
SDP
Catalog No. 5000241179
Encompass_Preferred
3-Azaspiro[5.5]undecane hydrochloride is a spirocyclic small molecule supplied as the hydrochloride salt for research use. It is a white to off-white solid with reported purity ≥97% and molecular weight 189.73 g·mol⁻¹. The compound has been reported to inhibit the wild-type influenza A virus M2 proton channel (A/M2) with an IC50 of 1 μM (two-electrode patch clamp assay).
  • Inhibits influenza A virus M2 proton channel (IC50 1 μM).
  • Supplied as a stable white to off-white solid.
  • Chemical formula C10H20ClN and molecular weight 189.73 g·mol⁻¹.
  • High reported purity (≥97%) suitable for research applications.
  • Available in multiple laboratory pack sizes for flexibility.
Catalog No. 50-002-41179 Supplier Medchemexpress LLC Supplier No. HY107755250MG
3-Azaspiro[5.5]undecane hydrochloride is a spirocyclic small molecule supplied as the hydrochloride salt for research use. It is a white to off-white solid with reported purity ≥97% and molecular weight 189.73 g·mol⁻¹. The compound has been reported to inhibit the wild-type influenza A virus M2 proton channel (A/M2) with an IC50 of 1 μM (two-electrode patch clamp assay).
  • Inhibits influenza A virus M2 proton channel (IC50 1 μM).
  • Supplied as a stable white to off-white solid.
  • Chemical formula C10H20ClN and molecular weight 189.73 g·mol⁻¹.
  • High reported purity (≥97%) suitable for research applications.
  • Available in multiple laboratory pack sizes for flexibility.