Learn More
Enzo Life Sciences 4,4-Pentamethylenepiperidine. HCl (50mg). CAS: 180-44-9
SDP
Supplier:  Enzo Life Sciences BMLCM1280050
Encompass_Preferred
A novel member of the spiro-piperidine class of M2 proton channel inhibitors similar to BL-1743. Displays increased potency (IC50=0.92µM) compared with amantadine. Alternative name: 3-Azaspiro[5.5]undecane . HCl. Purity: ≥97% (HPLC). Solubility: Soluble in water (>25mg/ml), DMSO (>25mg/ml) or 100% ethanol (>25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0435