Learn More
Medchemexpress LLC 3-Azetidinemethanol hydrochloride | 928038-44-2 | 123.58 | 500 MG
SDP
Catalog No. 5000400806
Encompass_Preferred
3-Azetidinemethanol hydrochloride is an intermediate that can be used in the synthesis of SHP2 inhibitor.
  • Can be used in the synthesis of SHP2 inhibitor.
  • Recommended storage conditions: 4°C, sealed storage, away from moisture.
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture).
  • Forms clear solutions in various solvents for in vitro and in vivo studies.
Catalog No. 50-004-00806 Supplier Medchemexpress LLC Supplier No. HY21336500MG
3-Azetidinemethanol hydrochloride is an intermediate that can be used in the synthesis of SHP2 inhibitor.
  • Can be used in the synthesis of SHP2 inhibitor.
  • Recommended storage conditions: 4°C, sealed storage, away from moisture.
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture).
  • Forms clear solutions in various solvents for in vitro and in vivo studies.