Learn More
Apexbio Technology LLC 3-Azetidinemethanol hydrochloride 928038-44-2 250mg
SDP
Catalog No. 5000093937
Encompass_Preferred
3-Azetidinemethanol hydrochloride (CAS 928038-44-2) is a nitrogen-containing heterocyclic compound commonly utilized as an intermediate in the synthesis of SHP2 inhibitors SHP2 a protein tyrosine phosphatase plays a regulatory role in cellular signaling pathways that control cell proliferation differentiation and migration Dysregulation of SHP2 function has been implicated in the development of various diseases including cancer By enabling the preparation of SHP2 inhibitors 3-Azetidinemethanol hydrochloride supports research aimed at elucidating the effects of SHP2 inhibition on cellular signaling pathways and exploring novel therapeutic strategies
Catalog No. 50-000-93937 Supplier Apexbio Technology LLC Supplier No. A4702250
3-Azetidinemethanol hydrochloride (CAS 928038-44-2) is a nitrogen-containing heterocyclic compound commonly utilized as an intermediate in the synthesis of SHP2 inhibitors SHP2 a protein tyrosine phosphatase plays a regulatory role in cellular signaling pathways that control cell proliferation differentiation and migration Dysregulation of SHP2 function has been implicated in the development of various diseases including cancer By enabling the preparation of SHP2 inhibitors 3-Azetidinemethanol hydrochloride supports research aimed at elucidating the effects of SHP2 inhibition on cellular signaling pathways and exploring novel therapeutic strategies