Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Azido-1-Propanamine

Catalog No. 762016100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-201-6100MG 100 mg
76-201-6250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 76-201-6100MG Supplier Sigma Aldrich Supplier No. 762016100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Azido-1-propanamine can be used to functionalize: Bismethylolpropionic acid (bis-MPA) monomers with azide functional group to generate high-generation dendrimers., Clickable zinc tetraphenylporphyrin scaffold with an azido group through click chemistry applicable in photodynamic therapy.3-Azido-1-propanamine may be used in the synthesis of mannopyranoside dendrimers for studying multivalent carbohydrate-protein interactions.

Specifications

CAS 88192-19-2
Density 1.020 g/mL at 25°C
Molecular Formula C3H8N4
Refractive Index n20/D 1.471
Linear Formula C3H8N4
MDL Number MFCD11046568
Quantity 100 mg
Synonym 3-Azidopropylamine
Molecular Weight (g/mol) 100.12 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.