Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Azido-1-Propanol

Catalog No. 7761305G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-613-05G 1 g
77-613-01G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 77-613-05G Supplier Sigma Aldrich Supplier No. 7761305G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Azido-1-propanol can be used as a building block for the synthesis of:Tris-(hydroxypropyltriazolylmethyl)amine (THPTA) which acts as a stabilizing ligand for Cu(I) to protect it from oxidation and disproportionation in click copolymerization.Heterofunctional polyesters by 1,3-dipolar cycloaddition reaction.Azide-containing cross-linkers.

Specifications

CAS 72320-38-8
Density 1.095 g/mL at 25°C
Molecular Formula C3H7N3O
Refractive Index n20/D 1.461
Linear Formula C3H7N3O
Quantity 1 g
MDL Number MFCD14652506
Synonym 1-Azidopropan-3-ol
Molecular Weight (g/mol) 101.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.