Learn More
Cayman Chemical 3'-Azido-2',3'-dideoxyuridine, Purity: ≥95%, CAS: 84472-85-5, 50 mg
SDP
Catalog No. 502914035
Encompass_Preferred
An antiviral nucleoside analog; inhibits cytopathogenicity induced by HIV or MSV in MT-4 cells (EC50s = 0.36 and 15 µM, respectively)
Catalog No. 50-291-4035 Supplier Cayman Chemical Supplier No. 3654450 MG
An antiviral nucleoside analog; inhibits cytopathogenicity induced by HIV or MSV in MT-4 cells (EC50s = 0.36 and 15 µM, respectively)