Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3′-Azido-2′,3′-dideoxyuridine | 84472-85-5 | 99.1% | 253.21 g·mol⁻¹ | C9H11N5O4 | 25 MG
SDP

Catalog No. 5000240815
Encompass_Preferred

3′-Azido-2′,3′-dideoxyuridine (AzdU, CAS 84472-85-5) is an azido-modified uridine nucleoside analog used in antiviral research and nucleotide chemistry. It acts as an inhibitor of HIV replication in cell-based assays and is provided as a solid for laboratory use.

  • Inhibits HIV replication in vitro.
  • High chemical purity (99.1%).
  • Molecular formula C9H11N5O4; molecular weight 253.21 g·mol⁻¹.
  • White to off-white solid suitable for synthesis and biochemical assays.
  • Powder stable at -20°C for long-term storage; solutions recommended at -80°C for extended storage.

Catalog No. 50-002-40815 Supplier Medchemexpress LLC Supplier No. HY10685025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3′-Azido-2′,3′-dideoxyuridine (AzdU, CAS 84472-85-5) is an azido-modified uridine nucleoside analog used in antiviral research and nucleotide chemistry. It acts as an inhibitor of HIV replication in cell-based assays and is provided as a solid for laboratory use.

  • Inhibits HIV replication in vitro.
  • High chemical purity (99.1%).
  • Molecular formula C9H11N5O4; molecular weight 253.21 g·mol⁻¹.
  • White to off-white solid suitable for synthesis and biochemical assays.
  • Powder stable at -20°C for long-term storage; solutions recommended at -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.