Learn More
Medchemexpress LLC 3'-azido-2',3'-dideoxyuridine | 84472-85-5 | MFCD00870324 | 99.1% | 253.21 g·mol⁻¹ | C9H11N5O4 | 50 MG
SDP
Catalog No. 5000242046
Encompass_Preferred
3′-Azido-2′,3′-dideoxyuridine (AzdU) is a nucleoside analog that inhibits human immunodeficiency virus replication and also functions as an azide-containing reagent for click-chemistry labeling. Supplied as a white to off-white solid with high purity and defined storage conditions, it is suitable for biochemical, virology, and chemical biology research.
  • Inhibits HIV replication in peripheral blood mononuclear cells.
  • Contains an azide group compatible with copper-catalyzed and strain-promoted click reactions.
  • High purity for analytical and biological assays.
  • Solid form with recommended storage for stability.
  • Suitable for nucleoside analog studies and chemical labeling applications.
Catalog No. 50-002-42046 Supplier Medchemexpress LLC Supplier No. HY10685050MG
3′-Azido-2′,3′-dideoxyuridine (AzdU) is a nucleoside analog that inhibits human immunodeficiency virus replication and also functions as an azide-containing reagent for click-chemistry labeling. Supplied as a white to off-white solid with high purity and defined storage conditions, it is suitable for biochemical, virology, and chemical biology research.
  • Inhibits HIV replication in peripheral blood mononuclear cells.
  • Contains an azide group compatible with copper-catalyzed and strain-promoted click reactions.
  • High purity for analytical and biological assays.
  • Solid form with recommended storage for stability.
  • Suitable for nucleoside analog studies and chemical labeling applications.