Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3'-azido-3'-deoxy-5-fluorocytidine | 2095417-18-6 | 99.9% | 286.22 g/mol | C9H11FN6O4 | 5 MG
SDP

Catalog No. 5000263674
Encompass_Preferred

3'-Azido-3'-deoxy-5-fluorocytidine is a cytidine derivative used as a click-chemistry reagent for bioorthogonal labeling and conjugation. It contains an azide group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC) reactions, and is supplied as a white to off-white solid with high reported purity.

  • Contains an azide functional group for bioconjugation.
  • Compatible with CuAAC and strain-promoted click reactions.
  • Reported purity 99.9%.
  • Soluble in DMSO (≈140 mg/mL).
  • Stable as a powder at -20 °C for long-term storage.
  • Available in small research quantities and solution formulations.

Catalog No. 50-002-63674 Supplier Medchemexpress LLC Supplier No. HY1116415MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3'-Azido-3'-deoxy-5-fluorocytidine is a cytidine derivative used as a click-chemistry reagent for bioorthogonal labeling and conjugation. It contains an azide group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC) reactions, and is supplied as a white to off-white solid with high reported purity.

  • Contains an azide functional group for bioconjugation.
  • Compatible with CuAAC and strain-promoted click reactions.
  • Reported purity 99.9%.
  • Soluble in DMSO (≈140 mg/mL).
  • Stable as a powder at -20 °C for long-term storage.
  • Available in small research quantities and solution formulations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.