Learn More
Medchemexpress LLC HY-11101 100mg Medchemexpress, Alendronate (sodium hydrate) CAS:121268-17-5 Purity:>98%
SDP
Catalog No. 501968811
Encompass_Preferred
Medchemexpress, HY-111640 50mg 3'-Azido-3'-deoxy-5-methylcytidine CAS:1282040-14-5 3'-Azido-3'-deoxy-5-methylcytidine (CS-92) is a potent xenotropic murine leukemia-related retrovirus (XMRV) inhibitor with a CC50 of 43.5 μM in MCF-7 cells. 3'-Azido-3'-deoxy-5-methylcytidine also inhibits HIV-1 reverse transcriptase with an EC50 of 0.06 μM in peripheral blood mononuclear (PBM) cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8811 Supplier Medchemexpress LLC Supplier No. HY11101100MG
Medchemexpress, HY-111640 50mg 3'-Azido-3'-deoxy-5-methylcytidine CAS:1282040-14-5 3'-Azido-3'-deoxy-5-methylcytidine (CS-92) is a potent xenotropic murine leukemia-related retrovirus (XMRV) inhibitor with a CC50 of 43.5 μM in MCF-7 cells. 3'-Azido-3'-deoxy-5-methylcytidine also inhibits HIV-1 reverse transcriptase with an EC50 of 0.06 μM in peripheral blood mononuclear (PBM) cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.