Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3'-azido-3'-deoxy-5-methylcytidine | 1282040-14-5 | 99.9% | 282.26 g/mol | C10H14N6O4 | 5 MG
SDP

Catalog No. 5000263675
Encompass_Preferred

3'-Azido-3'-deoxy-5-methylcytidine is an azide-functionalized nucleoside derivative used as an antiviral research reagent and a click-chemistry building block. The compound has reported inhibitory activity in cell-based antiviral assays and is suitable for copper-catalyzed and strain-promoted azide-alkyne cycloaddition bioconjugation.

  • Azide-functionalized nucleoside for bioconjugation via CuAAC and SPAAC.
  • Reported antiviral activity in cell-based assays, including XMRV and HIV-1 reverse transcriptase inhibition.
  • Molecular formula C10H14N6O4 and molecular weight 282.26 g/mol.
  • High purity (99.9%) suitable for research applications.
  • Light yellow to yellow solid supplied in small-mass laboratory packs.
  • Storage: powder at -20°C (long term) or 4°C; in solvent at -80°C or -20°C.

Catalog No. 50-002-63675 Supplier Medchemexpress LLC Supplier No. HY1116405MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3'-Azido-3'-deoxy-5-methylcytidine is an azide-functionalized nucleoside derivative used as an antiviral research reagent and a click-chemistry building block. The compound has reported inhibitory activity in cell-based antiviral assays and is suitable for copper-catalyzed and strain-promoted azide-alkyne cycloaddition bioconjugation.

  • Azide-functionalized nucleoside for bioconjugation via CuAAC and SPAAC.
  • Reported antiviral activity in cell-based assays, including XMRV and HIV-1 reverse transcriptase inhibition.
  • Molecular formula C10H14N6O4 and molecular weight 282.26 g/mol.
  • High purity (99.9%) suitable for research applications.
  • Light yellow to yellow solid supplied in small-mass laboratory packs.
  • Storage: powder at -20°C (long term) or 4°C; in solvent at -80°C or -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.