Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3'-Azido-3'-deoxy-beta-L-uridine | 2095417-28-8 | 98.8% | 269.21 | 1 ML
SDP

Catalog No. 5000438722
Encompass_Preferred

3'-Azido-3'-deoxy-beta-L-uridine (Compound 25) is a nucleoside derivative. It is a click chemistry reagent that contains an Azide group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.

  • Nucleoside derivative
  • Click chemistry reagent
  • Contains an Azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Appearance: Solid
  • Color: White to off-white
  • For research use only

Catalog No. 50-004-38722 Supplier Medchemexpress LLC Supplier No. HY11164210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3'-Azido-3'-deoxy-beta-L-uridine (Compound 25) is a nucleoside derivative. It is a click chemistry reagent that contains an Azide group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.

  • Nucleoside derivative
  • Click chemistry reagent
  • Contains an Azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Appearance: Solid
  • Color: White to off-white
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.