Learn More
Medchemexpress LLC 3'-Azido-3'-deoxy-beta-L-uridine | 2095417-28-8 | 98.8% | 269.21 | 100 MG
SDP
Catalog No. 5000439292
Encompass_Preferred
3'-Azido-3'-deoxy-beta-L-uridine is a nucleoside derivative that functions as a click chemistry reagent. It contains an azide group, allowing it to participate in various cycloaddition reactions.
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables participation in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing alkyne groups
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups
Catalog No. 50-004-39292 Supplier Medchemexpress LLC Supplier No. HY111642100MG
3'-Azido-3'-deoxy-beta-L-uridine is a nucleoside derivative that functions as a click chemistry reagent. It contains an azide group, allowing it to participate in various cycloaddition reactions.
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables participation in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing alkyne groups
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups