Learn More
Medchemexpress LLC 3'-azido-3'-deoxy-beta-L-uridine | 2095417-28-8 | MFCD30718534 | 98.8% | 269.21 g/mol | C9H11N5O5 | 5 MG
SDP
Catalog No. 5000263731
Encompass_Preferred
3-Azido-3-deoxy-beta-L-uridine is an azide-containing nucleoside derivative used as a clickable building block for bioconjugation and nucleoside chemistry. It participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC), enabling conjugation to alkyne- or cyclooctyne-functionalized molecules.
  • Azide functional group enabling CuAAC and SPAAC reactions.
  • Suitable for bioconjugation and nucleoside analogue synthesis.
  • High purity (98.8%) for reliable experimental use.
  • Molecular weight 269.21 g/mol; formula C9H11N5O5.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-63731 Supplier Medchemexpress LLC Supplier No. HY1116425MG
3-Azido-3-deoxy-beta-L-uridine is an azide-containing nucleoside derivative used as a clickable building block for bioconjugation and nucleoside chemistry. It participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC), enabling conjugation to alkyne- or cyclooctyne-functionalized molecules.
  • Azide functional group enabling CuAAC and SPAAC reactions.
  • Suitable for bioconjugation and nucleoside analogue synthesis.
  • High purity (98.8%) for reliable experimental use.
  • Molecular weight 269.21 g/mol; formula C9H11N5O5.
  • Available in small research pack sizes for laboratory use.