Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3'-azido-3'-deoxyadenosine | 58699-62-0 | MFCD27921039 | 98.4% | 292.25 g/mol | C10H12N8O3 | 5 MG
SDP

Catalog No. 5000263203
Encompass_Preferred

3'-Azido-3'-deoxyadenosine is a purine nucleoside analogue used in biochemical and biomedical research. It functions as a nucleoside antimetabolite with reported antitumor activity against indolent lymphoid malignancies and serves as an azide-containing reagent for bioorthogonal labeling via copper-catalyzed and strain-promoted azide-alkyne cycloadditions.

  • Purity: 98.44%.
  • Molecular formula: C10H12N8O3.
  • Molecular weight: 292.25 g/mol.
  • Appearance: white to off-white solid.
  • Storage: powder stable at -20°C for long-term storage; in solvent recommend -80°C short term.
  • Applications: nucleic acid metabolism studies, anticancer research, and click-chemistry labeling.

Catalog No. 50-002-63203 Supplier Medchemexpress LLC Supplier No. HY1318025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3'-Azido-3'-deoxyadenosine is a purine nucleoside analogue used in biochemical and biomedical research. It functions as a nucleoside antimetabolite with reported antitumor activity against indolent lymphoid malignancies and serves as an azide-containing reagent for bioorthogonal labeling via copper-catalyzed and strain-promoted azide-alkyne cycloadditions.

  • Purity: 98.44%.
  • Molecular formula: C10H12N8O3.
  • Molecular weight: 292.25 g/mol.
  • Appearance: white to off-white solid.
  • Storage: powder stable at -20°C for long-term storage; in solvent recommend -80°C short term.
  • Applications: nucleic acid metabolism studies, anticancer research, and click-chemistry labeling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.