Learn More
Medchemexpress LLC 3'-Azido-3'-deoxyuridine | 70580-88-0 | C9H11N5O5 | 5 MG
SDP
Catalog No. 5000372107
Encompass_Preferred
3'-Azido-3'-deoxyuridine is a purine nucleoside analogue with broad antitumor activity, particularly against indolent lymphoid malignancies. Its anticancer mechanisms involve inhibiting DNA synthesis and inducing apoptosis. This product is also a click chemistry reagent, containing an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • Purine nucleoside analogue
  • Exhibits broad antitumor activity
  • Inhibits DNA synthesis
  • Induces apoptosis
  • Click chemistry reagent with Azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-72107 Supplier Medchemexpress LLC Supplier No. HY1527205MG
3'-Azido-3'-deoxyuridine is a purine nucleoside analogue with broad antitumor activity, particularly against indolent lymphoid malignancies. Its anticancer mechanisms involve inhibiting DNA synthesis and inducing apoptosis. This product is also a click chemistry reagent, containing an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • Purine nucleoside analogue
  • Exhibits broad antitumor activity
  • Inhibits DNA synthesis
  • Induces apoptosis
  • Click chemistry reagent with Azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions