Learn More
Medchemexpress LLC 3-azido-D-alanine hydrochloride | 1379690-01-3 | C3H7ClN4O2 | 1 ML
SDP
Catalog No. 5000373139
Encompass_Preferred
3-Azido-D-alanine hydrochloride is an azido-modified D-alanine used in click chemistry labeling research. It contains an azide group, enabling its use in various cycloaddition reactions.
  • Used in click chemistry labeling research
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
  • Can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN group-containing molecules
Catalog No. 50-003-73139 Supplier Medchemexpress LLC Supplier No. HY151642A10MM/1ML
3-Azido-D-alanine hydrochloride is an azido-modified D-alanine used in click chemistry labeling research. It contains an azide group, enabling its use in various cycloaddition reactions.
  • Used in click chemistry labeling research
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
  • Can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN group-containing molecules