Learn More
Medchemexpress LLC 3-Azidopropanol | 72320-38-8 | C3H7N3O | 250 MG
SDP
Catalog No. 5000374537
Encompass_Preferred
3-Azidopropanol is an azide-containing alcohol and a click reaction substrate. It undergoes a copper-catalyzed addition reaction with propargyl alcohol.
  • Azide-containing alcohol
  • Click reaction substrate
  • High purity (99.52%)
  • Stable under recommended storage conditions
  • Colorless to light yellow liquid appearance
Catalog No. 50-003-74537 Supplier Medchemexpress LLC Supplier No. 250MGHY151851
3-Azidopropanol is an azide-containing alcohol and a click reaction substrate. It undergoes a copper-catalyzed addition reaction with propargyl alcohol.
  • Azide-containing alcohol
  • Click reaction substrate
  • High purity (99.52%)
  • Stable under recommended storage conditions
  • Colorless to light yellow liquid appearance