Learn More
Medchemexpress LLC 3-(benzo[b]thiophen-3-yl)-L-alanine | 72120-71-9 | MFCD00079684 | >98.5% | 221.28 g/mol | C11H11NO2S | 250 MG
SDP
Catalog No. 5000302824
Encompass_Preferred
(S)-2-Amino-3-(benzo[b]thiophen-3-yl)propanoic acid is the S-enantiomer of 3-benzothienylalanine, a chiral alanine derivative bearing a benzo[b]thiophene substituent. It is provided as a solid research reagent for use as a building block or reference standard in medicinal chemistry and biochemical studies.
  • Chiral S-enantiomer suitable for stereospecific synthesis and stereochemical studies.
  • Aromatic benzo[b]thiophene substituent at the 3-position.
  • Molecular formula C11H11NO2S and molecular weight 221.28 g/mol.
  • Supplied as a solid for laboratory use.
  • Appropriate as a building block or reference standard in medicinal chemistry workflows.
  • Store protected from light at 4°C; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-003-02824 Supplier Medchemexpress LLC Supplier No. 250MGHYW022796
(S)-2-Amino-3-(benzo[b]thiophen-3-yl)propanoic acid is the S-enantiomer of 3-benzothienylalanine, a chiral alanine derivative bearing a benzo[b]thiophene substituent. It is provided as a solid research reagent for use as a building block or reference standard in medicinal chemistry and biochemical studies.
  • Chiral S-enantiomer suitable for stereospecific synthesis and stereochemical studies.
  • Aromatic benzo[b]thiophene substituent at the 3-position.
  • Molecular formula C11H11NO2S and molecular weight 221.28 g/mol.
  • Supplied as a solid for laboratory use.
  • Appropriate as a building block or reference standard in medicinal chemistry workflows.
  • Store protected from light at 4°C; in solvent: -80°C (6 months), -20°C (1 month).